siRNA Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002004.2 | NP_001002004.2 | 16 | 2699 | ||
| GenBank mRNA | AK012059.1 | - | 3 | 970 | ||
| AK122206.1 | - | 1 | 351 | |||
| AK149719.1 | - | 16 | 2670 | |||
| BC040464.1 | - | 1 | 93 | |||
| BC075633.1 | - | 16 | 2659 | 30537398 | ||
| X81632.1 | - | 1 | 608 | |||

2610507B11Rik
Genomic Map