siRNA Details
Gene Symbol: Aldh1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028270.4 | NP_082546.1 | 2 | 391 | ||
GenBank mRNA | AK012213.1 | - | 2 | 286 | ||
AK088396.1 | - | 2 | 365 | |||
AK150992.1 | - | 2 | 293 | |||
AK151349.1 | - | 2 | 293 | |||
AK151364.1 | - | 2 | 293 | |||
AK153416.1 | - | 2 | 293 | |||
BC020001.1 | - | 2 | 295 | 4207479 | ||
BC086768.1 | - | 2 | 350 | 6808412 |
