siRNA Details
| Gene Symbol: Aldh1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028270.4 | NP_082546.1 | 2 | 652 | ||
| GenBank mRNA | AK012213.1 | - | 2 | 547 | ||
| AK088396.1 | - | 2 | 626 | |||
| AK150992.1 | - | 2 | 554 | |||
| AK151349.1 | - | 2 | 554 | |||
| AK151364.1 | - | 2 | 554 | |||
| AK153416.1 | - | 2 | 554 | |||
| BC020001.1 | - | 2 | 556 | 4207479 | ||
| BC086768.1 | - | 2 | 611 | 6808412 | ||

Aldh1b1
Genomic Map