siRNA Details
| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028276.1 | NP_082552.1 | 9 | 838 | ||
| GenBank mRNA | AK134348.1 | - | 9 | 896 | ||
| AK134784.1 | - | 9 | 863 | |||
| AK163230.1 | - | 1 | 944 | |||
| AK172924.1 | - | 9 | 838 | |||
| AY316163.1 | - | 9 | 861 | |||
| BC062165.1 | - | 9 | 887 | |||
| BC082590.1 | - | 9 | 864 | 30625504 | ||

Utp14a
Genomic Map