siRNA Details
| Gene Symbol: Naalad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028279.3 | NP_082555.2 | 2 | 160 | ||
| GenBank mRNA | AK133611.1 | - | 2 | 270 | ||
| AK135653.1 | - | 4 | 302 | |||
| AK161457.1 | - | 5 | 466 | |||
| AY243507.1 | - | 2 | 160 | |||
| BC108995.1 | - | 2 | 144 | 40047859 | ||
| BC108996.1 | - | 2 | 144 | 40047860 | ||

Naalad2
Genomic Map