siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139140.1 | NP_631879.1 | 13 | 1838 | ||
| GenBank mRNA | AB079132.1 | - | 13 | 1838 | ||
| AK012237.1 | - | 2 | 219 | |||
| AK159335.1 | - | 13 | 1933 | |||
| AK167788.1 | - | 12 | 1516 | |||
| BC023432.1 | - | 11 | 1488 | 5029821 | ||
| BC072579.1 | - | 13 | 2105 | |||

Spats2
Genomic Map