siRNA Details
| Gene Symbol: Car10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028296.3 | NP_082572.1 | 7 | 1586 | ||
| GenBank mRNA | AB080741.1 | - | 8 | 1542 | ||
| AK132222.1 | - | 7 | 1427 | |||
| BC017606.1 | - | 7 | 778 | 4507552 | ||
| BC066111.1 | - | 8 | 1470 | 30360743 | ||

Car10
Genomic Map