siRNA Details
| Gene Symbol: Tmem209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178625.3 | NP_848740.3 | 14 | 1604 | ||
| GenBank mRNA | AK029366.1 | - | 11 | 1293 | ||
| AK044877.1 | - | 14 | 1620 | |||
| AK078544.1 | - | 14 | 1618 | |||
| AK089486.1 | - | 1479 | ||||
| AK089962.1 | - | 14 | 1619 | |||
| AK170465.1 | - | 1479 | ||||
| BC052187.1 | - | 14 | 1647 | 6393580 | ||
| BC057109.1 | - | 14 | 1604 | 6400080 | ||

Tmem209
Genomic Map