siRNA Details
| Gene Symbol: Hnrnpll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144802.4 | NP_659051.3 | 8 | 1321 | ||
| GenBank mRNA | AK012866.1 | - | 8 | 1037 | ||
| AK146879.1 | - | 8 | 1283 | |||
| AK160943.1 | - | 7 | 1129 | |||
| BC004763.1 | - | 5 | 294 | |||
| BC012849.1 | - | 8 | 1267 | 3587716 | ||

Hnrnpll
Genomic Map