siRNA Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028341.4 | NP_082617.2 | 6 | 1239 | ||
| GenBank mRNA | AK013267.1 | - | 6 | 974 | ||
| AK077971.1 | - | 299 | ||||
| BC020021.1 | - | 6 | 989 | 2648772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc39c| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028341.4 | NP_082617.2 | 6 | 1239 | ||
| GenBank mRNA | AK013267.1 | - | 6 | 974 | ||
| AK077971.1 | - | 299 | ||||
| BC020021.1 | - | 6 | 989 | 2648772 | ||