siRNA Details
| Gene Symbol: Tmem135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028343.4 | NP_082619.3 | 2 | 447 | ||
| GenBank mRNA | AK013269.1 | - | 2 | 421 | ||
| AK028401.1 | - | 2 | 257 | |||
| AK028846.1 | - | 2 | 421 | |||
| AK047200.1 | - | 2 | 421 | |||
| AK153175.1 | - | 2 | 371 | |||
| AY040841.1 | - | 2 | 297 | |||
| BC033279.1 | - | 2 | 427 | 4165990 | ||
| BC050931.1 | - | 2 | 356 | 6811187 | ||

Tmem135
Genomic Map