siRNA Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289568.1 | NP_001276497.1 | 2 | 234 | ||
| NM_028349.3 | NP_082625.2 | 2 | 234 | |||
| GenBank mRNA | AK013338.1 | - | 2, 3 | 209 | ||
| AK165224.1 | - | 2 | 180 | |||
| BB617231.1 | - | 2 | 236 | |||
| BC050110.1 | - | 2 | 184 | 30024366 | ||

Sass6
Genomic Map