siRNA Details
Gene Symbol: Mon1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028369.3 | NP_082645.1 | 4 | 1627 | ||
GenBank mRNA | AK013387.1 | - | 4 | 1574 | ||
AK172624.1 | - | 4 | 1534 | |||
AK211446.1 | - | 1 | 59 | |||
BC006070.1 | - | 1 | 434 | |||
BC048852.1 | - | 4 | 1553 | 5148425 | ||
BC058717.1 | - | 4 | 1605 | 5704178 |
