siRNA Details
| Gene Symbol: Setd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028385.1 | NP_082661.1 | 14 | 2261 | ||
| GenBank mRNA | AK049143.1 | - | 8 | 1339 | ||
| AK122551.1 | - | 14 | 2261 | |||
| BC056502.1 | - | 17 | 2600 | |||
| BC083184.1 | - | 13 | 2142 | 30536831 | ||

Setd5
Genomic Map