siRNA Details
| Gene Symbol: Setd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028385.1 | NP_082661.1 | 22 | 4059 | ||
| GenBank mRNA | AK053541.1 | - | 7 | 2190 | ||
| AK122551.1 | - | 22 | 4059 | |||
| BC083184.1 | - | 21 | 3940 | 30536831 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Setd5| Gene Symbol: Setd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028385.1 | NP_082661.1 | 22 | 4059 | ||
| GenBank mRNA | AK053541.1 | - | 7 | 2190 | ||
| AK122551.1 | - | 22 | 4059 | |||
| BC083184.1 | - | 21 | 3940 | 30536831 | ||