siRNA Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252620.1 | NP_001239549.1 | 9 | 840 | ||
| NM_001252621.1 | NP_001239550.1 | 8 | 831 | |||
| NM_023689.3 | NP_076178.1 | 9 | 954 | |||
| GenBank mRNA | AJ278998.1 | - | 9 | 848 | ||
| AK013644.1 | - | 9 | 883 | |||
| AK049843.1 | - | 8 | 832 | |||
| AK140736.1 | - | 2 | 141 | |||
| AK220280.1 | - | 9 | 938 | |||
| BC017601.1 | - | 9 | 853 | 4502169 | ||
| BC053334.1 | - | 9 | 825 | 6492113 | ||

Spock3
Genomic Map