siRNA Details
| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028410.1 | NP_082686.1 | 5 | 889 | ||
| GenBank mRNA | AK013663.1 | - | 5 | 980 | ||
| BC049103.1 | - | 5 | 889 | 6400126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkrir| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028410.1 | NP_082686.1 | 5 | 889 | ||
| GenBank mRNA | AK013663.1 | - | 5 | 980 | ||
| BC049103.1 | - | 5 | 889 | 6400126 | ||