siRNA Details
| Gene Symbol: Appl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145221.2 | NP_660256.1 | 17 | 1650 | ||
| GenBank mRNA | AK030445.1 | - | 17 | 1529 | ||
| AK033566.1 | - | 17 | 1621 | |||
| AK045438.1 | - | 17 | 1586 | |||
| AK049307.1 | - | 17 | 1637 | |||
| AK146356.1 | - | 17 | 1621 | |||
| AK173169.1 | - | 2 | 101 | |||
| AY113705.1 | - | 17 | 1615 | |||
| BC019708.1 | - | 9 | 928 | |||
| BC063751.1 | - | 17 | 1662 | 6511401 | ||

Appl1
Genomic Map