siRNA Details
| Gene Symbol: Appl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145221.2 | NP_660256.1 | 10 | 884 | ||
| GenBank mRNA | AK030445.1 | - | 10 | 764 | ||
| AK033566.1 | - | 10 | 855 | |||
| AK045438.1 | - | 10 | 820 | |||
| AK049307.1 | - | 10 | 871 | |||
| AK146356.1 | - | 10 | 855 | |||
| AY113705.1 | - | 10 | 849 | |||
| BC019708.1 | - | 2 | 162 | |||
| BC063751.1 | - | 10 | 896 | 6511401 | ||

Appl1
Genomic Map