siRNA Details
| Gene Symbol: Insig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271531.1 | NP_001258460.1 | 4 | 705 | ||
| NM_001271532.1 | NP_001258461.1 | 3 | 572 | |||
| NM_133748.2 | NP_598509.1 | 4 | 948 | |||
| NM_178082.3 | NP_835183.1 | 4 | 712 | |||
| GenBank mRNA | AF527631.1 | - | 4 | 682 | ||
| AK040703.1 | - | 4 | 703 | |||
| AK050394.1 | - | 4 | 706 | |||
| AK152076.1 | - | 4 | 678 | |||
| AK154543.1 | - | 4 | 711 | |||
| AK161256.1 | - | 4 | 716 | |||
| AK165634.1 | - | 4 | 717 | |||
| AK190310.1 | - | 2 | 183 | |||
| AK191083.1 | - | 2 | 199 | |||
| AK192853.1 | - | 2 | 157 | |||
| BC015288.1 | - | 4 | 662 | 4240711 | ||
| BC023067.1 | - | 4 | 686 | 5357599 | ||
| BC023874.1 | - | 3 | 604 | 5355039 | ||
| BC024411.1 | - | 4 | 700 | 4971297 | ||

Insig2
Genomic Map