siRNA Details
Gene Symbol: Gpr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175191.4 | NP_780400.2 | 3 | 1922 | ||
GenBank mRNA | AK034194.1 | - | 3 | 1876 | ||
AK048077.1 | - | 3 | 1877 | |||
AK141569.1 | - | 3 | 1907 | |||
AY255527.1 | - | 1 | 326 | |||
BC037625.1 | - | 1 | 376 | |||
BC089509.1 | - | 3 | 1923 | 30252116 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
