siRNA Details
| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028427.3 | NP_082703.1 | 4 | 681 | ||
| GenBank mRNA | AK013998.1 | - | 4 | 680 | ||
| AK046181.1 | - | 4 | 653 | |||
| AK132619.1 | - | 4 | 652 | |||
| AK168146.1 | - | 4 | 664 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem192| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028427.3 | NP_082703.1 | 4 | 681 | ||
| GenBank mRNA | AK013998.1 | - | 4 | 680 | ||
| AK046181.1 | - | 4 | 653 | |||
| AK132619.1 | - | 4 | 652 | |||
| AK168146.1 | - | 4 | 664 | |||