siRNA Details
| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163747.1 | NP_001157219.1 | 4, 5 | 674 | ||
| NM_028427.3 | NP_082703.1 | 5, 6 | 809 | |||
| GenBank mRNA | AI849438.1 | - | 1 | 328 | ||
| AK013998.1 | - | 5, 6 | 808 | |||
| AK046181.1 | - | 5, 6 | 781 | |||
| AK168146.1 | - | 5, 6 | 792 | |||
| BC020157.1 | - | 4, 5 | 605 | 3708193 | ||

Tmem192
Genomic Map