siRNA Details
| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163747.1 | NP_001157219.1 | 3 | 505 | ||
| NM_028427.3 | NP_082703.1 | 3 | 505 | |||
| GenBank mRNA | AK013998.1 | - | 3 | 504 | ||
| AK046181.1 | - | 3 | 477 | |||
| AK132619.1 | - | 3 | 472 | |||
| AK168146.1 | - | 3 | 488 | |||
| BC020157.1 | - | 3 | 436 | 3708193 | ||

Tmem192
Genomic Map