siRNA Details
| Gene Symbol: Prr36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207203.2 | NP_997086.2 | 5 | 908 | ||
| GenBank mRNA | BC068157.1 | - | 5 | 908 | 30363411 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr36| Gene Symbol: Prr36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207203.2 | NP_997086.2 | 5 | 908 | ||
| GenBank mRNA | BC068157.1 | - | 5 | 908 | 30363411 | |