siRNA Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153587.2 | NP_705815.1 | 14 | 1912 | ||
| GenBank mRNA | AK030051.1 | - | 12 | 1658 | ||
| AK032316.1 | - | 13 | 1810 | |||
| AK046319.1 | - | 13 | 1494 | |||
| AK160758.1 | - | 6 | 676 | |||
| AY341873.1 | - | 15 | 1901 | |||
| AY341881.1 | - | 13 | 1706 | |||
| BC035298.1 | - | 15 | 1912 | 4021865 | ||

Rps6ka5
Genomic Map