siRNA Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153587.2 | NP_705815.1 | 13 | 1649 | ||
| GenBank mRNA | AY341873.1 | - | 14 | 1638 | ||
| BC035298.1 | - | 14 | 1649 | 4021865 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6ka5| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153587.2 | NP_705815.1 | 13 | 1649 | ||
| GenBank mRNA | AY341873.1 | - | 14 | 1638 | ||
| BC035298.1 | - | 14 | 1649 | 4021865 | ||