siRNA Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153587.2 | NP_705815.1 | 17, 18 | 2499 | ||
| GenBank mRNA | AK030051.1 | - | 15, 16 | 2245 | ||
| AK032316.1 | - | 16, 17 | 2397 | |||
| AK160758.1 | - | 9, 10 | 1263 | |||
| AY341873.1 | - | 18, 19 | 2488 | |||
| AY341881.1 | - | 16, 17 | 2293 | |||
| BC035298.1 | - | 18, 19 | 2499 | 4021865 | ||
| BC038372.1 | - | 3, 4 | 2404 | |||

Rps6ka5
Genomic Map