siRNA Details
| Gene Symbol: Golim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291069.1 | NP_001277998.1 | 15, 16 | 2558 | ||
| NM_175193.6 | NP_780402.1 | 14, 15 | 2474 | |||
| GenBank mRNA | AK033730.1 | - | 14, 15 | 2462 | ||
| AK048434.1 | - | 15, 16 | 2385 | |||
| AK050346.1 | - | 2, 3 | 142 | |||
| AK078928.1 | - | 15, 16 | 2558 | |||
| BC023824.1 | - | 1, 2 | 376 | |||
| BC086642.1 | - | 14, 15 | 2306 | 6836803 | ||
| BC089354.1 | - | 14, 15 | 2356 | 6837211 | ||

Golim4
Genomic Map