siRNA Details
| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028447.3 | NP_082723.2 | 6 | 946 | ||
| GenBank mRNA | AK143537.1 | - | 6 | 938 | ||
| BC014817.1 | - | 6 | 862 | 3709357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrc1| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028447.3 | NP_082723.2 | 6 | 946 | ||
| GenBank mRNA | AK143537.1 | - | 6 | 938 | ||
| BC014817.1 | - | 6 | 862 | 3709357 | ||