siRNA Details
| Gene Symbol: Tm7sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028454.2 | NP_082730.2 | 7 | 880 | ||
| GenBank mRNA | AF480070.1 | - | 7 | 754 | ||
| AK014166.1 | - | 7 | 861 | |||
| AK155475.1 | - | 7 | 837 | |||
| BC014769.1 | - | 7 | 872 | 4239159 | ||

Tm7sf2
Genomic Map