siRNA Details
| Gene Symbol: 3110043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081343.1 | NP_001074812.1 | 5 | 803 | ||
| GenBank mRNA | AK014175.1 | - | 4 | 255 | ||
| AK154817.1 | - | 5 | 777 | |||
| AK167354.1 | - | 5 | 734 | |||
| BC026738.1 | - | 3 | 208 | 4020832 | ||
| BC076612.1 | - | 5 | 905 | 30638238 | ||

3110043O21Rik
Genomic Map