siRNA Details
| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028474.1 | NP_082750.1 | 4 | 2288 | ||
| GenBank mRNA | BC151166.1 | - | 3 | 1980 | 9088055 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptchd4| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028474.1 | NP_082750.1 | 4 | 2288 | ||
| GenBank mRNA | BC151166.1 | - | 3 | 1980 | 9088055 | |