siRNA Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028478.2 | NP_082754.2 | 8 | 838 | ||
| GenBank mRNA | AK005472.1 | - | 8 | 786 | ||
| AK132288.1 | - | 8 | 800 | |||
| BC031808.1 | - | 8 | 810 | |||
| BC039934.1 | - | 8 | 885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf6| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028478.2 | NP_082754.2 | 8 | 838 | ||
| GenBank mRNA | AK005472.1 | - | 8 | 786 | ||
| AK132288.1 | - | 8 | 800 | |||
| BC031808.1 | - | 8 | 810 | |||
| BC039934.1 | - | 8 | 885 | |||