siRNA Details
| Gene Symbol: Rhobtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028493.2 | NP_082769.1 | 7 | 1408 | ||
| GenBank mRNA | AK006650.1 | - | 3 | 527 | ||
| AK129234.1 | - | 7 | 1451 | |||
| AK139158.1 | - | 7 | 1288 | |||
| BC050836.1 | - | 7 | 1408 | 6394317 | ||

Rhobtb3
Genomic Map