siRNA Details
| Gene Symbol: Phospho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028521.2 | NP_082797.1 | 4 | 699 | ||
| GenBank mRNA | AK006724.1 | - | 4 | 700 | ||
| AK132362.1 | - | 2 | 1827 | |||
| AK159551.1 | - | 4 | 746 | |||
| BC025612.1 | - | 4 | 735 | 5257820 | ||
| BC031523.1 | - | 2 | 370 | 4009719 | ||

Phospho2
Genomic Map