siRNA Details
| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175199.3 | NP_780408.1 | 4 | 376 | ||
| GenBank mRNA | AB093239.1 | - | 4 | 320 | ||
| AK006867.1 | - | 4 | 360 | |||
| AK038491.1 | - | 4 | 348 | |||
| AK087732.1 | - | 5 | 533 | |||
| AK141925.1 | - | 4 | 377 | |||
| AK165073.1 | - | 4 | 404 | |||
| AY196789.1 | - | 4 | 375 | |||
| BC030362.1 | - | 4 | 346 | 5369051 | ||

Hspa12a
Genomic Map