siRNA Details
| Gene Symbol: 1700067K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183097.2 | NP_898920.2 | 4 | 497 | ||
| GenBank mRNA | AK006917.1 | - | 4 | 766 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700067K01Rik| Gene Symbol: 1700067K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183097.2 | NP_898920.2 | 4 | 497 | ||
| GenBank mRNA | AK006917.1 | - | 4 | 766 | ||