siRNA Details
Gene Symbol: 1700067K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183097.2 | NP_898920.2 | 3 | 373 | ||
GenBank mRNA | AK006917.1 | - | 3 | 642 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700067K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183097.2 | NP_898920.2 | 3 | 373 | ||
GenBank mRNA | AK006917.1 | - | 3 | 642 |