siRNA Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175200.4 | NP_780409.2 | 5 | 740 | ||
| GenBank mRNA | AK076617.1 | - | 5 | 742 | ||
| BC118929.2 | - | 5 | 706 | 40090362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Als2cr11| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175200.4 | NP_780409.2 | 5 | 740 | ||
| GenBank mRNA | AK076617.1 | - | 5 | 742 | ||
| BC118929.2 | - | 5 | 706 | 40090362 | ||