siRNA Details
| Gene Symbol: Rnf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038993.3 | NP_001034082.1 | 10 | 1347 | ||
| NM_175201.5 | NP_780410.2 | 9 | 1377 | |||
| GenBank mRNA | AK147608.1 | - | 10 | 1404 | ||
| AK163346.1 | - | 5 | 508 | |||
| BC060730.1 | - | 9 | 1354 | 6408623 | ||
| BC062976.1 | - | 9 | 1291 | 6848803 | ||

Rnf38
Genomic Map