siRNA Details
| Gene Symbol: Rnf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038993.3 | NP_001034082.1 | 6 | 907 | ||
| NM_175201.5 | NP_780410.2 | 5 | 937 | |||
| GenBank mRNA | AK006888.1 | - | 5 | 898 | ||
| AK147608.1 | - | 6 | 964 | |||
| BC060730.1 | - | 5 | 914 | 6408623 | ||
| BC062976.1 | - | 5 | 851 | 6848803 | ||

Rnf38
Genomic Map