siRNA Details
| Gene Symbol: Camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028576.2 | NP_082852.1 | 2 | 183 | ||
| GenBank mRNA | AK007130.1 | - | 2 | 278 | ||
| AK155246.1 | - | 2 | 184 | |||
| AK172547.1 | - | 2 | 129 | |||
| BC138272.1 | - | 2 | 331 | 8861291 | ||
| BC138273.1 | - | 2 | 331 | 8861292 | ||
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK147783.1 | - | 220 | |||

Camkmt
Genomic Map