siRNA Details
| Gene Symbol: Cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028593.2 | NP_082869.2 | 4 | 775 | ||
| GenBank mRNA | AF354666.1 | - | 4 | 717 | ||
| AK008849.1 | - | 4 | 721 | |||
| AK029112.1 | - | 4 | 778 | |||
| BC027665.1 | - | 2 | 271 | |||
| BC147289.1 | - | 2 | 263 | |||
| BC147290.1 | - | 2 | 263 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AB023478.1 | - | 316 | Chr. - - [-] on Build | ||

Cybrd1
Genomic Map