siRNA Details
| Gene Symbol: Cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028593.2 | NP_082869.2 | 1, 2 | 269 | ||
| GenBank mRNA | AF354666.1 | - | 1, 2 | 211 | ||
| AK008849.1 | - | 1, 2 | 215 | |||
| AK029112.1 | - | 1, 2 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cybrd1| Gene Symbol: Cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028593.2 | NP_082869.2 | 1, 2 | 269 | ||
| GenBank mRNA | AF354666.1 | - | 1, 2 | 211 | ||
| AK008849.1 | - | 1, 2 | 215 | |||
| AK029112.1 | - | 1, 2 | 272 | |||