siRNA Details
| Gene Symbol: Ttc21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047604.2 | NP_001041069.1 | 9, 10 | 1178 | ||
| NM_001290669.1 | NP_001277598.1 | 9, 10 | 1178 | |||
| GenBank mRNA | AK010712.1 | - | 6, 7 | 682 | ||
| AK147810.1 | - | 2, 3 | 261 | |||
| BC151199.1 | - | 9, 10 | 1153 | |||
| BC172115.1 | - | 9, 10 | 1153 | |||
| DQ011164.1 | - | 9, 10 | 1178 | |||

Ttc21b
Genomic Map