siRNA Details
| Gene Symbol: Zbtb8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028603.4 | NP_082879.1 | 3 | 657 | ||
| GenBank mRNA | AK010736.1 | - | 3 | 656 | ||
| AK135238.1 | - | 1 | 1011 | |||
| BC017614.1 | - | 3 | 488 | 4912757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb8a| Gene Symbol: Zbtb8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028603.4 | NP_082879.1 | 3 | 657 | ||
| GenBank mRNA | AK010736.1 | - | 3 | 656 | ||
| AK135238.1 | - | 1 | 1011 | |||
| BC017614.1 | - | 3 | 488 | 4912757 | ||