siRNA Details
Gene Symbol: Rbm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170847.1 | NP_001164318.1 | 11 | 3308 | ||
GenBank mRNA | BC038663.1 | - | 3 | 1344 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170847.1 | NP_001164318.1 | 11 | 3308 | ||
GenBank mRNA | BC038663.1 | - | 3 | 1344 |