siRNA Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028658.4 | NP_082934.3 | 15 | 1652 | ||
| GenBank mRNA | AK028185.1 | - | 15 | 1516 | ||
| AK155094.1 | - | 15 | 1484 | |||
| AK170254.1 | - | 15 | 1540 | |||
| BC137673.1 | - | 15 | 1592 | 8860692 | ||

Ppp1r21
Genomic Map