siRNA Details
| Gene Symbol: Psmb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175204.4 | NP_780413.1 | 1 | 892 | ||
| GenBank mRNA | AB299436.1 | - | 1 | 875 | ||
| AK017902.1 | - | 1 | 894 | |||
| AK037368.1 | - | 1 | 890 | |||
| AK041393.1 | - | 1 | 890 | |||
| AK162727.1 | - | 1 | 894 | |||

Psmb11
Genomic Map